Cyclopropyl-2-(4-methoxyphenyl)ethyl ketone ≥97%

Supplier: Matrix Scientific

086856-1G 086856-2G 086856-5G
102851-038EA 672.31 USD
102851-038 102851-040 102851-042
Cyclopropyl-2-(4-methoxyphenyl)ethyl ketone ≥97%
Cyclopropyl-2-(4-methoxyphenyl)ethyl ketone

Hazard: Irritant

Formula: C₁₃H₁₆O₂
MDL Number: MFCD03843193
CAS Number: 87141-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR