2',4'-Dichloro-3-(3-methoxyphenyl)propiophenone ≥97%

Supplier: Matrix Scientific

086855-1G 086855-2G 086855-5G
102851-032EA 672.31 USD
102851-032 102851-034 102851-036
2',4'-Dichloro-3-(3-methoxyphenyl)propiophenone ≥97%
2',4'-Dichloro-3-(3-methoxyphenyl)propiophenone

Hazard: Irritant

Formula: C₁₆H₁₄Cl₂O₂
MDL Number: MFCD03843135
CAS Number: 898775-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR