Cyclobutyl-2-(4-methylphenyl)ethyl ketone ≥97%

Supplier: Matrix Scientific

086853-1G 086853-2G 086853-5G
102851-020EA 672.31 USD
102851-020 102851-022 102851-024
Cyclobutyl-2-(4-methylphenyl)ethyl ketone ≥97%
Cyclobutyl-2-(4-methylphenyl)ethyl ketone

Hazard: Irritant

Formula: C₁₄H₁₈O
MDL Number: MFCD03843041
CAS Number: 898769-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR