Cyclopropyl-2-(4-methylphenyl)ethyl ketone ≥97%

Supplier: Matrix Scientific

086852-1G 086852-2G 086852-5G
102851-014EA 672.31 USD
102851-014 102851-016 102851-018
Cyclopropyl-2-(4-methylphenyl)ethyl ketone ≥97%
Cyclopropyl-2-(4-methylphenyl)ethyl ketone

Hazard: Irritant

Formula: C₁₃H₁₆O
MDL Number: MFCD03843040
CAS Number: 898769-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR