2'-Carboethoxy-3-(4-methylphenyl)propiophenone ≥97%

Supplier: Matrix Scientific

086851-1G 086851-2G 086851-5G
102851-008EA 672.31 USD
102851-008 102851-010 102851-012
2'-Carboethoxy-3-(4-methylphenyl)propiophenone ≥97%
2'-Carboethoxy-3-(4-methylphenyl)propiophenone

Hazard: Irritant

Formula: C₁₉H₂₀O₃
MDL Number: MFCD03843039
CAS Number: 898768-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR