Cyclopropyl-2-(3-methylphenyl)ethyl ketone ≥97%

Supplier: Matrix Scientific

086850-1G 086850-2G 086850-5G
102851-002EA 672.31 USD
102851-002 102851-004 102851-006
Cyclopropyl-2-(3-methylphenyl)ethyl ketone ≥97%
Cyclopropyl-2-(3-methylphenyl)ethyl ketone

Hazard: Irritant

Formula: C₁₃H₁₆O
MDL Number: MFCD03843025
CAS Number: 898768-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR