Cyclopropyl-2-(2-methylphenyl)ethyl ketone ≥97%

Supplier: Matrix Scientific

086849-1G 086849-2G 086849-5G
102850-996EA 672.31 USD
102850-996 102850-998 102851-000
Cyclopropyl-2-(2-methylphenyl)ethyl ketone ≥97%
Cyclopropyl-2-(2-methylphenyl)ethyl ketone

Hazard: Irritant

Formula: C₁₃H₁₆O
MDL Number: MFCD03843009
CAS Number: 898790-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR