Cyclohexyl-2-phenethyl ketone ≥97%

Supplier: Matrix Scientific

086848-1G 086848-2G 086848-5G
102850-990EA 672.31 USD
102850-990 102850-992 102850-994
Cyclohexyl-2-phenethyl ketone ≥97%
Cyclohexyl-2-phenethyl ketone

Hazard: Irritant

Formula: C₁₅H₂₀O
MDL Number: MFCD00965990
CAS Number: 43125-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR