Cyclobutyl-2-phenethyl ketone ≥97%

Supplier: Matrix Scientific

086846-1G 086846-2G 086846-5G
102850-978EA 672.31 USD
102850-978 102850-980 102850-982
Cyclobutyl-2-phenethyl ketone ≥97%
Cyclobutyl-2-phenethyl ketone

Hazard: Irritant

Formula: C₁₃H₁₆O
MDL Number: MFCD03842959
CAS Number: 874009-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR