4'-Fluoro-3-phenylpropiophenone ≥97%

Supplier: Matrix Scientific

086845-1G 086845-2G 086845-5G
102850-972EA 672.31 USD
102850-972 102850-974 102850-976
4'-Fluoro-3-phenylpropiophenone ≥97%
4'-Fluoro-3-phenylpropiophenone

Hazard: Irritant

Formula: C₁₅H₁₃FO
MDL Number: MFCD03842930
CAS Number: 41938-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR