3'-Carboethoxy-3-phenylpropiophenone ≥97%

Supplier: Matrix Scientific

086843-1G 086843-2G 086843-5G
102850-960EA 672.31 USD
102850-960 102850-962 102850-964
3'-Carboethoxy-3-phenylpropiophenone ≥97%
3'-Carboethoxy-3-phenylpropiophenone

Hazard: Irritant

Formula: C₁₈H₁₈O₃
MDL Number: MFCD03842922
CAS Number: 898764-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR