2'-Carboethoxy-3-phenylpropiophenone ≥97%

Supplier: Matrix Scientific

086842-1G 086842-2G 086842-5G
102850-954EA 672.31 USD
102850-954 102850-956 102850-958
2'-Carboethoxy-3-phenylpropiophenone ≥97%
2'-Carboethoxy-3-phenylpropiophenone

Hazard: Irritant

Formula: C₁₈H₁₈O₃
CAS Number: 898764-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR