2'-Methyl-3-phenylpropiophenone ≥97%

Supplier: Matrix Scientific

086840-1G 086840-2G 086840-5G
102850-942EA 672.31 USD
102850-942 102850-944 102850-946
2'-Methyl-3-phenylpropiophenone ≥97%
2'-Methyl-3-phenylpropiophenone

Hazard: Irritant

Formula: C₁₆H₁₆O
MDL Number: MFCD03842914
CAS Number: 93433-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR