2-[8-(1,4-Dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone ≥97%

Supplier: Matrix Scientific

086838-1G 086838-2G 086838-5G
102850-930EA 672.31 USD
102850-930 102850-932 102850-934
2-[8-(1,4-Dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone ≥97%
2-[8-(1,4-Dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

Hazard: Irritant

Formula: C₂₁H₂₃NO₃
MDL Number: MFCD03842687
CAS Number: 898755-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR