Cyclohexyl-2-(morpholinomethyl)phenyl ketone ≥97%
Supplier: Matrix Scientific
086837-1G
086837-2G
086837-5G
102850-924EA
672.31
USD
102850-924
102850-926
102850-928
Cyclohexyl-2-(morpholinomethyl)phenyl ketone ≥97%
Cyclohexyl-2-(morpholinomethyl)phenyl ketone
Hazard: Irritant
Formula:
C₁₈H₂₅NO₂ |
MDL Number:
MFCD03842453 CAS Number: 898751-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...