Ethyl-4-[4-(4-methylpiperazinomethyl)phenyl]-4-oxobutyrate ≥97%

Supplier: Matrix Scientific

086836-1G 086836-2G 086836-5G
102850-918EA 672.31 USD
102850-918 102850-920 102850-922
Ethyl-4-[4-(4-methylpiperazinomethyl)phenyl]-4-oxobutyrate ≥97%
Ethyl-4-[4-(4-methylpiperazinomethyl)phenyl]-4-oxobutyrate

Hazard: Irritant

Formula: C₁₈H₂₆N₂O₃
MW: 318.42 g/mol
MDL Number: MFCD03841831
CAS Number: 898763-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR