4-Chloro-4'-pyrrolidinomethyl benzophenone ≥97%

Supplier: Matrix Scientific

086835-1G 086835-2G 086835-5G
102850-912EA 672.31 USD
102850-912 102850-914 102850-916
4-Chloro-4'-pyrrolidinomethyl benzophenone ≥97%
4-Chloro-4'-pyrrolidinomethyl benzophenone

Hazard: Irritant

Formula: C₁₈H₁₈ClNO
MDL Number: MFCD03841513
CAS Number: 742085-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR