Ethyl-6-[4-(morpholinomethyl)phenyl]-6-oxohexanoate ≥97%

Supplier: Matrix Scientific

086834-1G 086834-2G 086834-5G
102850-906EA 672.31 USD
102850-906 102850-908 102850-910
Ethyl-6-[4-(morpholinomethyl)phenyl]-6-oxohexanoate ≥97%
Ethyl-6-[4-(morpholinomethyl)phenyl]-6-oxohexanoate

Hazard: Irritant

Formula: C₁₉H₂₇NO₄
MDL Number: MFCD03841435
CAS Number: 898770-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR