Cyclohexyl-3,5-dimethylphenyl ketone ≥97%

Supplier: Matrix Scientific

086832-1G 086832-2G 086832-5G
102850-894EA 672.31 USD
102850-894 102850-896 102850-898
Cyclohexyl-3,5-dimethylphenyl ketone ≥97%
Cyclohexyl-3,5-dimethylphenyl ketone

Hazard: Irritant

Formula: C₁₅H₂₀O
MDL Number: MFCD03841359
CAS Number: 898769-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR