Cyclohexyl-3,4-dimethylphenyl ketone ≥97%

Supplier: Matrix Scientific

086831-1G 086831-2G 086831-5G
102850-888EA 672.31 USD
102850-888 102850-890 102850-892
Cyclohexyl-3,4-dimethylphenyl ketone ≥97%
Cyclohexyl-3,4-dimethylphenyl ketone

Hazard: Irritant

Formula: C₁₅H₂₀O
MDL Number: MFCD03841358
CAS Number: 133047-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR