Cyclohexyl-2,6-dimethylphenyl ketone ≥97%

Supplier: Matrix Scientific

086830-1G 086830-2G 086830-5G
102850-882EA 672.31 USD
102850-882 102850-884 102850-886
Cyclohexyl-2,6-dimethylphenyl ketone ≥97%
Cyclohexyl-2,6-dimethylphenyl ketone

Hazard: Irritant

Formula: C₁₅H₂₀O
MDL Number: MFCD03841357
CAS Number: 898769-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR