Cyclohexyl-2,5-dimethylphenyl ketone ≥97%

Supplier: Matrix Scientific

086829-1G 086829-2G 086829-5G
102850-876EA 672.31 USD
102850-876 102850-878 102850-880
Cyclohexyl-2,5-dimethylphenyl ketone ≥97%
Cyclohexyl-2,5-dimethylphenyl ketone

Hazard: Irritant

Formula: C₁₅H₂₀O
MDL Number: MFCD00447621
CAS Number: 2890-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR