Cyclohexyl-2,4-dimethylphenyl ketone ≥97%

Supplier: Matrix Scientific

086828-1G 086828-2G 086828-5G
102850-870EA 672.31 USD
102850-870 102850-872 102850-874
Cyclohexyl-2,4-dimethylphenyl ketone ≥97%
Cyclohexyl-2,4-dimethylphenyl ketone

Hazard: Irritant

Formula: C₁₅H₂₀O
MDL Number: MFCD03841356
CAS Number: 2760-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR