4-Chlorophenyl cyclohexyl ketone ≥97%

Supplier: Matrix Scientific

086826-1G 086826-2G 086826-5G
102850-858EA 672.31 USD
102850-858 102850-860 102850-862
4-Chlorophenyl cyclohexyl ketone ≥97%
4-Chlorophenyl cyclohexyl ketone

Hazard: Irritant

Formula: C₁₃H₁₅ClO
MDL Number: MFCD00454416
CAS Number: 3277-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR