4-Bromophenyl cyclohexyl ketone ≥97%

Supplier: Matrix Scientific

086825-1G 086825-2G 086825-5G
102850-852EA 672.31 USD
102850-852 102850-854 102850-856
4-Bromophenyl cyclohexyl ketone ≥97%
4-Bromophenyl cyclohexyl ketone

Hazard: Irritant

Formula: C₁₃H₁₅BrO
MDL Number: MFCD03841351
CAS Number: 3277-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR