Cyclohexyl-4-thiomethylphenyl ketone ≥97%

Supplier: Matrix Scientific

086824-1G 086824-2G 086824-5G
102850-846EA 672.31 USD
102850-846 102850-848 102850-850
Cyclohexyl-4-thiomethylphenyl ketone ≥97%
Cyclohexyl-4-thiomethylphenyl ketone

Hazard: Irritant

Formula: C₁₄H₁₈OS
MDL Number: MFCD03841349
CAS Number: 197439-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR