Cyclohexyl-2-thiomethylphenyl ketone ≥97%

Supplier: Matrix Scientific

086823-1G 086823-2G 086823-5G
102850-840EA 672.31 USD
102850-840 102850-842 102850-844
Cyclohexyl-2-thiomethylphenyl ketone ≥97%
Cyclohexyl-2-thiomethylphenyl ketone

Hazard: Irritant

Formula: C₁₄H₁₈OS
MDL Number: MFCD03841348
CAS Number: 898769-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR