4-Cyanophenyl cyclohexyl ketone ≥97%

Supplier: Matrix Scientific

086822-1G 086822-2G 086822-5G
102850-834EA 672.31 USD
102850-834 102850-836 102850-838
4-Cyanophenyl cyclohexyl ketone ≥97%
4-Cyanophenyl cyclohexyl ketone

Hazard: Irritant

Formula: C₁₄H₁₅NO
MDL Number: MFCD03841344
CAS Number: 898792-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR