Cyclohexyl-4-methoxyphenyl ketone ≥97%

Supplier: Matrix Scientific

086821-1G 086821-2G 086821-5G
102850-828EA 672.31 USD
102850-828 102850-830 102850-832
Cyclohexyl-4-methoxyphenyl ketone ≥97%
Cyclohexyl-4-methoxyphenyl ketone

Hazard: Irritant

Formula: C₁₄H₁₈O₂
MW: 218.3 g/mol
MDL Number: MFCD03841341
CAS Number: 7469-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR