Cyclohexyl-3-methoxyphenyl ketone ≥97%

Supplier: Matrix Scientific

086820-1G 086820-2G 086820-5G
102850-822EA 672.31 USD
102850-822 102850-824 102850-826
Cyclohexyl-3-methoxyphenyl ketone ≥97%
Cyclohexyl-3-methoxyphenyl ketone

Hazard: Irritant

Formula: C₁₄H₁₈O₂
MDL Number: MFCD03841340
CAS Number: 898792-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR