Cyclohexyl-2-methoxyphenyl ketone ≥97%

Supplier: Matrix Scientific

086819-1G 086819-2G 086819-5G
102850-816EA 672.31 USD
102850-816 102850-818 102850-820
Cyclohexyl-2-methoxyphenyl ketone ≥97%
Cyclohexyl-2-methoxyphenyl ketone

Hazard: Irritant

Formula: C₁₄H₁₈O₂
MDL Number: MFCD03841339
CAS Number: 111504-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR