Cyclohexyl-4-methylphenyl ketone ≥97%

Supplier: Matrix Scientific

086818-1G 086818-2G 086818-5G
102850-810EA 672.31 USD
102850-810 102850-812 102850-814
Cyclohexyl-4-methylphenyl ketone ≥97%
Cyclohexyl-4-methylphenyl ketone

Hazard: Irritant

Formula: C₁₄H₁₈O
MDL Number: MFCD01871291
CAS Number: 2789-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR