Cyclohexyl-3-methylphenyl ketone ≥97%

Supplier: Matrix Scientific

Synonyms: Cyclohexyl(m-tolyl)methanone

086817-1G 086817-2G 086817-5G
102850-804EA 672.31 USD
102850-804 102850-806 102850-808
Cyclohexyl-3-methylphenyl ketone ≥97%
Cyclohexyl-3-methylphenyl ketone

Hazard: Irritant

Formula: C₁₄H₁₈O
MW: 202.3 g/mol
MDL Number: MFCD03841338
CAS Number: 3277-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR