2-Fluorophenyl cyclopentyl ketone ≥97%

Supplier: Matrix Scientific

086815-1G 086815-2G 086815-5G
102850-792EA 672.31 USD
102850-792 102850-794 102850-796
2-Fluorophenyl cyclopentyl ketone ≥97%
2-Fluorophenyl cyclopentyl ketone

Hazard: Irritant

Formula: C₁₂H₁₃FO
MDL Number: MFCD03841320
CAS Number: 111982-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR