4-Bromo-3-fluorophenyl cyclopentyl ketone ≥97%
Supplier: Matrix Scientific
086814-1G
086814-2G
086814-5G
102850-786EA
672.31
USD
102850-786
102850-788
102850-790
4-Bromo-3-fluorophenyl cyclopentyl ketone ≥97%
4-Bromo-3-fluorophenyl cyclopentyl ketone
Hazard: Irritant
Formula:
C₁₂H₁₂BrFO |
MDL Number:
MFCD03841317 CAS Number: 898791-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...