4-Bromophenyl cyclopentyl ketone ≥97%
Supplier: Matrix Scientific
086813-1G
086813-2G
086813-5G
102850-780EA
672.31
USD
102850-780
102850-782
102850-784
4-Bromophenyl cyclopentyl ketone ≥97%
4-Bromophenyl cyclopentyl ketone
Hazard: Irritant
Formula:
C₁₂H₁₃BrO |
MDL Number:
MFCD03841306 CAS Number: 2204-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...