3-Bromophenyl cyclopentyl ketone ≥97%
Supplier: Matrix Scientific
086812-1G
086812-2G
086812-5G
102850-774EA
672.31
USD
102850-774
102850-776
102850-778
3-Bromophenyl cyclopentyl ketone ≥97%
3-Bromophenyl cyclopentyl ketone
Hazard: Irritant
Formula:
C₁₂H₁₃BrO |
MDL Number:
MFCD03841305 CAS Number: 898791-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...