3-Carboethoxyphenyl cyclopentyl ketone ≥97%

Supplier: Matrix Scientific

086811-1G 086811-2G 086811-5G
102850-768EA 672.31 USD
102850-768 102850-770 102850-772
3-Carboethoxyphenyl cyclopentyl ketone ≥97%
3-Carboethoxyphenyl cyclopentyl ketone

Hazard: Irritant

Formula: C₁₅H₁₈O₃
MDL Number: MFCD03841301
CAS Number: 684271-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR