3-Cyanophenyl cyclopentyl ketone ≥97%

Supplier: Matrix Scientific

086810-1G 086810-2G 086810-5G
102850-762EA 672.31 USD
102850-762 102850-764 102850-766
3-Cyanophenyl cyclopentyl ketone ≥97%
3-Cyanophenyl cyclopentyl ketone

Hazard: Irritant

Formula: C₁₃H₁₃NO
MDL Number: MFCD03841298
CAS Number: 898791-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR