Cyclopentyl-4-methoxyphenyl ketone ≥97%

Supplier: Matrix Scientific

086809-1G 086809-2G
102850-756EA 672.31 USD
102850-756 102850-758
Cyclopentyl-4-methoxyphenyl ketone ≥97%
Cyclopentyl-4-methoxyphenyl ketone

Hazard: Irritant

Formula: C₁₃H₁₆O₂
MDL Number: MFCD03841296
CAS Number: 15831-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR