Cyclopentyl-4-methylphenyl ketone ≥97%

Supplier: Matrix Scientific

086808-1G 086808-2G 086808-5G
102850-750EA 672.31 USD
102850-750 102850-752 102850-754
Cyclopentyl-4-methylphenyl ketone ≥97%
Cyclopentyl-4-methylphenyl ketone

Hazard: Irritant

Formula: C₁₃H₁₆O
MDL Number: MFCD03841293
CAS Number: 97802-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR