Cyclopentyl-2-methylphenyl ketone ≥97%
Supplier: Matrix Scientific
086806-1G
086806-2G
086806-5G
102850-738EA
672.31
USD
102850-738
102850-740
102850-742
Cyclopentyl-2-methylphenyl ketone ≥97%
Cyclopentyl-2-methylphenyl ketone
Hazard: Irritant
Formula:
C₁₃H₁₆O |
MDL Number:
MFCD03841291 CAS Number: 7063-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...