2-Fluorophenyl cyclobutyl ketone ≥97%

Supplier: Matrix Scientific

086805-1G 086805-2G 086805-5G
102850-732EA 672.31 USD
102850-732 102850-734 102850-736
2-Fluorophenyl cyclobutyl ketone ≥97%
2-Fluorophenyl cyclobutyl ketone

Hazard: Irritant

Formula: C₁₁H₁₁FO
MDL Number: MFCD03841274
CAS Number: 898790-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR