4-Bromo-3-fluorophenyl cyclobutyl ketone ≥97%
Supplier: Matrix Scientific
086803-1G
086803-2G
086803-5G
102850-720EA
672.31
USD
102850-720
102850-722
102850-724
4-Bromo-3-fluorophenyl cyclobutyl ketone ≥97%
4-Bromo-3-fluorophenyl cyclobutyl ketone
Hazard: Irritant
Formula:
C₁₁H₁₀BrFO |
MDL Number:
MFCD03841270 CAS Number: 898790-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...