Cyclobutyl-2,6-dimethylphenyl ketone ≥97%

Supplier: Matrix Scientific

086801-1G 086801-2G 086801-5G
102850-708EA 672.31 USD
102850-708 102850-710 102850-712
Cyclobutyl-2,6-dimethylphenyl ketone ≥97%
Cyclobutyl-2,6-dimethylphenyl ketone

Hazard: Irritant

Formula: C₁₃H₁₆O
MDL Number: MFCD03841267
CAS Number: 898790-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR