4-Chlorophenyl cyclobutyl ketone ≥97%
Supplier: Matrix Scientific
086800-1G
086800-2G
086800-5G
102850-702EA
672.31
USD
102850-702
102850-704
102850-706
4-Chlorophenyl cyclobutyl ketone ≥97%
4-Chlorophenyl cyclobutyl ketone
Hazard: Irritant
Formula:
C₁₁H₁₁ClO |
MDL Number:
MFCD03841262 CAS Number: 77585-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...