4-Carboethoxyphenyl cyclobutyl ketone ≥97%
Supplier: Matrix Scientific
086798-1G
086798-2G
086798-5G
102850-690EA
672.31
USD
102850-690
102850-692
102850-694
4-Carboethoxyphenyl cyclobutyl ketone ≥97%
4-Carboethoxyphenyl cyclobutyl ketone
Hazard: Irritant
Formula:
C₁₄H₁₆O₃ |
MDL Number:
MFCD03841256 CAS Number: 801303-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...