2-Carboethoxyphenyl cyclobutyl ketone ≥97%
Supplier: Matrix Scientific
086797-1G
086797-2G
086797-5G
102850-684EA
672.31
USD
102850-684
102850-686
102850-688
2-Carboethoxyphenyl cyclobutyl ketone ≥97%
2-Carboethoxyphenyl cyclobutyl ketone
Hazard: Irritant
Formula:
C₁₄H₁₆O₃ |
MDL Number:
MFCD03841254 CAS Number: 898790-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...