2-Cyanophenyl cyclobutyl ketone ≥97%
Supplier: Matrix Scientific
086796-1G
086796-2G
086796-5G
102850-678EA
672.31
USD
102850-678
102850-680
102850-682
2-Cyanophenyl cyclobutyl ketone ≥97%
2-Cyanophenyl cyclobutyl ketone
Hazard: Irritant
Formula:
C₁₂H₁₁NO |
MDL Number:
MFCD03841251 CAS Number: 898790-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...