Cyclobutyl-4-methoxyphenyl ketone ≥97%

Supplier: Matrix Scientific

086795-1G 086795-2G 086795-5G
102850-672EA 856.92 USD
102850-672 102850-674 102850-676
Cyclobutyl-4-methoxyphenyl ketone ≥97%
Cyclobutyl-4-methoxyphenyl ketone

Hazard: Irritant

Formula: C₁₂H₁₄O₂
MDL Number: MFCD03841250
CAS Number: 100121-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR